Search
Search

Mob1b| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046278.1 | - | 1945 | 102 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
|---|
Set Membership: |
|