Search
Search

Try10| Gene Symbol: Try10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038996.2 | NP_001034085.1 | 1 | 50 | 74 | |
| GenBank mRNA | BC138698.1 | - | 1 | 50 | 8861720 | 74 |
| BC146017.1 | - | 1 | 50 | 40131320 | 74 | |
Set Membership: |
|