Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm99999051_gH
          See other CCR2 GE Assays ›
          Gene Symbol:
          Ccr2
          Gene Name:
          chemokine (C-C motif) receptor 2
          Gene Aliases:
          Cc-ckr-2, Ccr2a, Ccr2b, Ckr2, Ckr2a, Ckr2b, Cmkbr2, mJe-r
          Chromosome Location:
          Chr.9: 124101918 - 124109140 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm99999051_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009915.2 NP_034045.1 3 443 60
          GenBank mRNA AK046579.1 - 677 60
          AK138599.1 - 2 756 60
          BC138803.1 - 2 403 8861825 60
          BC138804.1 - 2 403 8861826 60
          U47035.1 - 2 352 60
          U51717.1 - 2 401 60
          U56819.1 - 3 443 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Chemokine Panel Mouse Immune Response Inventoried Mouse_Immune_Response Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccr2

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel remodeling
          wound healing involved in inflammatory response
          monocyte chemotaxis
          positive regulation of T-helper 1 type immune response
          negative regulation of type 2 immune response
          chemotaxis
          inflammatory response
          immune response
          humoral immune response
          cellular defense response
          signal transduction
          G-protein coupled receptor signaling pathway
          regulation of vascular endothelial growth factor production
          positive regulation of T cell chemotaxis
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          sensory perception of pain
          cellular homeostasis
          hemopoiesis
          regulation of cell migration
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          T-helper 17 cell chemotaxis
          positive regulation of tumor necrosis factor biosynthetic process
          apoptotic cell clearance
          negative regulation of eosinophil degranulation
          positive regulation of T-helper 1 cell differentiation
          positive regulation of alpha-beta T cell proliferation
          positive regulation of inflammatory response
          positive regulation of T cell activation
          chemokine-mediated signaling pathway
          positive regulation of monocyte chemotaxis
          positive regulation of immune complex clearance by monocytes and macrophages
          positive regulation of monocyte extravasation
          positive regulation of CD8-positive, alpha-beta T cell extravasation
          positive regulation of astrocyte chemotaxis
          positive regulation of hematopoietic stem cell migration
          signal transducer activity
          G-protein coupled receptor activity
          chemokine receptor activity
          protein binding
          C-C chemokine receptor activity
          cytokine binding
          chemokine (C-C motif) ligand 2 binding
          chemokine (C-C motif) ligand 12 binding
          chemokine (C-C motif) ligand 7 binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245989 U56819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245989 U56819.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24207 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120315 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137202 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245989 U56819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245989 U56819.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245989 U56819.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245989 U56819.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06958231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120315 - -
          Whole Mouse Genome G4122A A_52_P580853 AK162705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580853 NM_009915.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245989 U56819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245989 U56819.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112634 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120315 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26898248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07782382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08176109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08072106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07739685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07713083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08176099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08056629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08024330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08093937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07909374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26898246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590633 - -
          Affymetrix HT MG-430 PM Array Plate 1421187_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421186_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460067_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421188_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590631 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598013 - -
          Affymetrix Mouse Gene 1.1 ST Array 10590635 - -
          Mouse Genome 430 2.0 Array 1421187_PM_at - -
          Mouse Genome 430 2.0 Array 1421188_PM_at - -
          Mouse Genome 430 2.0 Array 1421187_at - -
          Mouse Genome 430 2.0 Array 1460067_PM_at - -
          Mouse Genome 430 2.0 Array 1421186_at - -
          Mouse Genome 430 2.0 Array 1460067_at - -
          Mouse Genome 430 2.0 Array 1421188_at - -
          Mouse Genome 430 2.0 Array 1421186_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12772_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12772_at - -
          Affymetrix Mouse Clariom S Array PSR0900014073.mm.1 NM_009915.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014073.mm.1 NM_009915.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014073.mm.1 TC0900001674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014074.mm.1 NM_009915.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014074.mm.1 NM_009915.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014074.mm.1 TC0900001674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014076.mm.1 NM_009915.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014076.mm.1 NM_009915.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014076.mm.1 TC0900001674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014071.mm.1 NM_009915.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014071.mm.1 NM_009915.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014071.mm.1 TC0900001674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014075.mm.1 NM_009915.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014075.mm.1 NM_009915.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014075.mm.1 TC0900001674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014068.mm.1 NM_009915.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014068.mm.1 NM_009915.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014068.mm.1 TC0900001674.mm.1 MTA TC
          Mouse Expression Set 430 1421188_PM_at - -
          Mouse Expression Set 430 1421187_at - -
          Mouse Expression Set 430 1421187_PM_at - -
          Mouse Expression Set 430 1421186_at - -
          Mouse Expression Set 430 1421186_PM_at - -
          Mouse Expression Set 430 1421188_at - -
          Mouse Expression Set 430 1460067_PM_at - -
          Mouse Expression Set 430 1460067_at - -
          Murine Genome U74v2 Set 93397_PM_at - -
          Murine Genome U74v2 Set 93397_at - -
          Mouse Genome 430A 2.0 Array 1421187_at - -
          Mouse Genome 430A 2.0 Array 1421186_PM_at - -
          Mouse Genome 430A 2.0 Array 1421188_at - -
          Mouse Genome 430A 2.0 Array 1421187_PM_at - -
          Mouse Genome 430A 2.0 Array 1421186_at - -
          Mouse Genome 430A 2.0 Array 1421188_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421187_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421187_at - -
          Affymetrix HT Mouse Genome 430A Array 1421188_at - -
          Affymetrix HT Mouse Genome 430A Array 1421188_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421186_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421186_at - -
          Affymetrix HT Mouse Genome 430B Array 1460067_at - -
          Affymetrix HT Mouse Genome 430B Array 1460067_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450519 - -
          Illumina mouse-6 v1.0 expression beadchip 107000601 - -
          Illumina mouse-6 v1.0 expression beadchip 1450519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222795 - -
          mouse-6 v1.1 (Illumina) scl36282.3_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450519 - -
          Illumina Sentrix mouse-6.v1 bead array 107000601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36282.3_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107000601 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226563 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222795 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline