Search
Search

METTL16| Gene Symbol: METTL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835870.1 | XP_028691703.1 | 891 | 111 | ||
| XM_028835871.1 | XP_028691704.1 | 726 | 111 | |||
| XM_028835872.1 | XP_028691705.1 | 738 | 111 | |||
| GenBank mRNA | JU493269.1 | - | 855 | 111 | ||
| JV661183.1 | - | 857 | 111 | |||
Set Membership: |
|