Search
Search

MTCP1| Gene Symbol: MTCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194094.1 | NP_001181023.1 | 268 | 83 | ||
| XM_015128571.1 | XP_014984057.1 | 153 | 83 | |||
| GenBank mRNA | JU472130.1 | - | 168 | 83 | ||
| JV652057.1 | - | 103 | 83 | |||
Set Membership: |
|