Search
Search

ADORA1| Gene Symbol: ADORA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261667.1 | NP_001248596.1 | 145 | 104 | ||
| XM_015118614.2 | XP_014974100.1 | 364 | 104 | |||
| XM_028839954.1 | XP_028695787.1 | 125 | 104 | |||
| XM_028840003.1 | XP_028695836.1 | 349 | 104 | |||
| GenBank mRNA | JU332965.1 | - | 151 | 104 | ||
| JV046217.1 | - | 145 | 104 | |||
Set Membership: |
|