Search
Search

COX7B2| Gene Symbol: COX7B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193385.2 | NP_001180314.1 | 53 | 78 | ||
| XM_028847920.1 | XP_028703753.1 | 77 | 78 | |||
| GenBank mRNA | JV635991.1 | - | 162 | 78 | ||
Set Membership: |
|