Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562837_m1
          See other AQP5 GE Assays ›
          Gene Symbol:
          Aqp5
          Gene Name:
          aquaporin 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 141249044 - 141252576 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aqp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012779.1 NP_036911.1 1 - 2 473 69
          GenBank mRNA BC078904.1 - 1 - 2 467 69
          BC092572.1 - 1 - 2 462 7122974 69
          BC110045.1 - 1 - 2 478 7100092 69
          U16245.1 - 1 - 2 473 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001818.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aqp5

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          water transport
          cellular water homeostasis
          carbon dioxide transport
          glycerol transport
          pancreatic juice secretion
          ion transmembrane transport
          odontogenesis
          saliva secretion
          camera-type eye morphogenesis
          water channel activity
          glycerol channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147325 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11560 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11560 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11560 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700594448 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62364 - -
          Agilent Rat cDNA Microarray (G4105A) 700594448 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899155 - -
          Affymetrix Rat Gene 1.0 ST Array 10899156 - -
          Affymetrix Rat Gene 1.0 ST Array 10899154 - -
          Affymetrix Rat Gene 1.0 ST Array 10899152 - -
          Affymetrix Rat Gene 1.0 ST Array 10899153 - -
          Affymetrix Rat Gene 1.1 ST Array 10899151 - -
          Rat Genome 230 2.0 Array 1368469_PM_at - -
          Rat Genome 230 2.0 Array 1368469_at - -
          Affymetrix HT Rat Focus Array 1368469_at - -
          Affymetrix HT Rat Focus Array 1368469_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700014023.rn.1 TC0700001818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014021.rn.1 TC0700001818.rn.1 RTA TC
          Rat Expression Set 230 1368469_at - -
          Rat Expression Set 230 1368469_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899151 - -
          Rat Genome U34 Set U16245_g_at - -
          Rat Genome U34 Set U16245_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22264937 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364664 - -
          RatRef-12 Expression BeadChip ILMN_49974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline