Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569203_m1
          See other ISL1 GE Assays ›
          Gene Symbol:
          Isl1
          Gene Name:
          ISL LIM homeobox 1
          Gene Aliases:
          Isl-1, isl-1=homeobox
          Chromosome Location:
          Chr.2: 48488911 - 48501217 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569203_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Isl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017339.3 NP_059035.3 3 - 4 485 106
          GenBank mRNA AY557632.1 - 3 - 4 480 106
          S69329.1 - 3 - 4 485 106
          X53258.1 - 3 - 4 588 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002949.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Isl1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural crest cell migration
          heart morphogenesis
          secondary heart field specification
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          endocardial cushion morphogenesis
          cardiac right ventricle morphogenesis
          regulation of secondary heart field cardioblast proliferation
          regulation of transcription, DNA-templated
          heart development
          positive regulation of cell proliferation
          regulation of gene expression
          positive regulation of vascular endothelial growth factor production
          spinal cord motor neuron cell fate specification
          spinal cord motor neuron differentiation
          visceral motor neuron differentiation
          trigeminal nerve development
          pituitary gland development
          neuron differentiation
          pancreas development
          axon regeneration
          retinal ganglion cell axon guidance
          positive regulation of insulin secretion
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 alpha production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          negative regulation of intracellular estrogen receptor signaling pathway
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          negative regulation of neuron differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          neuron fate commitment
          neuron fate specification
          mesenchymal cell differentiation
          sensory system development
          peripheral nervous system neuron development
          peripheral nervous system neuron axonogenesis
          negative regulation of inflammatory response
          ventricular cardiac muscle tissue morphogenesis
          pharyngeal system development
          cardiac muscle cell myoblast differentiation
          innervation
          atrial septum morphogenesis
          cardiac cell fate determination
          cellular response to glucocorticoid stimulus
          positive regulation of granulocyte colony-stimulating factor production
          negative regulation of protein homodimerization activity
          negative regulation of canonical Wnt signaling pathway
          positive regulation of macrophage colony-stimulating factor production
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          enhancer sequence-specific DNA binding
          DNA binding
          chromatin binding
          protein binding
          zinc ion binding
          ligand-dependent nuclear receptor binding
          estrogen receptor binding
          bHLH transcription factor binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P509917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P509917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P509917 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P509917 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P509917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34819 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821487 - -
          Affymetrix Rat Gene 1.0 ST Array 10821490 - -
          Affymetrix Rat Gene 1.0 ST Array 10821489 - -
          Affymetrix Rat Gene 1.0 ST Array 10821488 - -
          Affymetrix Rat Gene 1.0 ST Array 10821492 - -
          Affymetrix Rat Gene 1.0 ST Array 10821491 - -
          Affymetrix Rat Gene 1.1 ST Array 10821486 - -
          Rat Genome 230 2.0 Array 1369681_at - -
          Rat Genome 230 2.0 Array 1369681_PM_at - -
          Rat Genome 230 2.0 Array 1396895_at - -
          Rat Genome 230 2.0 Array 1396895_PM_at - -
          Affymetrix HT Rat Focus Array 1396895_at - -
          Affymetrix HT Rat Focus Array 1369681_at - -
          Affymetrix HT Rat Focus Array 1369681_PM_at - -
          Affymetrix HT Rat Focus Array 1396895_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200020745.rn.1 TC0200002949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020742.rn.1 TC0200002949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020744.rn.1 TC0200002949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020749.rn.1 TC0200002949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020750.rn.1 TC0200002949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020748.rn.1 TC0200002949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200020746.rn.1 TC0200002949.rn.1 RTA TC
          Rat Expression Set 230 1369681_at - -
          Rat Expression Set 230 1369681_PM_at - -
          Rat Expression Set 230 1396895_PM_at - -
          Rat Expression Set 230 1396895_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821486 - -
          Rat Genome U34 Set S69329_g_at - -
          Rat Genome U34 Set S69329_at - -
          Rat Genome U34 Set rc_AA944599_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21414494 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371866 - -
          RatRef-12 Expression BeadChip ILMN_69643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline