Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574768_m1
          See other PPAP2A GE Assays ›
          Gene Symbol:
          Ppap2a
          Gene Name:
          phosphatidic acid phosphatase type 2A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 44664085 - 44726820 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574768_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppap2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022538.2 NP_071983.2 1 - 2 153 97
          GenBank mRNA BC061815.1 - 1 - 2 153 5598693 97
          JQ013728.1 - 1 - 2 91 97
          U90556.1 - 1 - 2 68 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000395.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppap2a

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          lipid metabolic process
          dephosphorylation
          phosphatidate phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047832 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134737 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047832 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047832 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047832 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33526 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84471 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60143 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813057 - -
          Affymetrix Rat Gene 1.0 ST Array 10813053 - -
          Affymetrix Rat Gene 1.0 ST Array 10813055 - -
          Affymetrix Rat Gene 1.0 ST Array 10813056 - -
          Affymetrix Rat Gene 1.0 ST Array 10813049 - -
          Affymetrix Rat Gene 1.0 ST Array 10813052 - -
          Affymetrix Rat Gene 1.0 ST Array 10813050 - -
          Affymetrix Rat Gene 1.0 ST Array 10813054 - -
          Affymetrix Rat Gene 1.1 ST Array 10813048 - -
          Rat Genome 230 2.0 Array 1369961_PM_at - -
          Rat Genome 230 2.0 Array 1369961_at - -
          Rat Genome 230 2.0 Array 1378893_at - -
          Rat Genome 230 2.0 Array 1378893_PM_at - -
          Affymetrix HT Rat Focus Array 1369961_PM_at - -
          Affymetrix HT Rat Focus Array 1369961_at - -
          Affymetrix Rat Clariom S Assay PSR0200002366.rn.1 TC0200000395.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002365.rn.1 TC0200000395.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002359.rn.1 TC0200000395.rn.1 RTA TC
          Rat Expression Set 230 1369961_PM_at - -
          Rat Expression Set 230 1369961_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813048 - -
          Rat Genome U34 Set rc_AA818593_g_at - -
          Rat Genome U34 Set rc_AA818593_at - -
          Rat Genome U34 Set rc_AI230354_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21800532 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650461 - -
          RatRef-12 Expression BeadChip ILMN_56065 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline