Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00583468_m1
          See other CHN2 GE Assays ›
          Gene Symbol:
          Chn2
          Gene Name:
          chimerin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 84423708 - 84452989 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00583468_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032084.1 NP_114473.1 6 - 7 635 130
          GenBank mRNA L07494.1 - 6 - 7 635 130
          X69489.1 - 6 - 7 635 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000947.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chn2

          Gene Ontology Categories:

          Function(s) Process(es)

          acrosome assembly
          intracellular signal transduction
          phorbol ester receptor activity
          GTPase activator activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P419291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P419291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P419291 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P419291 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P419291 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701611831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59486 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36638 - -
          Agilent Rat cDNA Microarray (G4105A) 701611831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855630 - -
          Affymetrix Rat Gene 1.0 ST Array 10855625 - -
          Affymetrix Rat Gene 1.0 ST Array 10855628 - -
          Affymetrix Rat Gene 1.0 ST Array 10855629 - -
          Affymetrix Rat Gene 1.0 ST Array 10855631 - -
          Affymetrix Rat Gene 1.0 ST Array 10855627 - -
          Affymetrix Rat Gene 1.0 ST Array 10855623 - -
          Affymetrix Rat Gene 1.0 ST Array 10855626 - -
          Affymetrix Rat Gene 1.0 ST Array 10855622 - -
          Affymetrix Rat Gene 1.1 ST Array 10855615 - -
          Rat Genome 230 2.0 Array 1376800_PM_at - -
          Rat Genome 230 2.0 Array 1368675_PM_at - -
          Rat Genome 230 2.0 Array 1394833_PM_at - -
          Rat Genome 230 2.0 Array 1394833_at - -
          Rat Genome 230 2.0 Array 1368675_at - -
          Rat Genome 230 2.0 Array 1376800_at - -
          Affymetrix HT Rat Focus Array 1368675_at - -
          Affymetrix HT Rat Focus Array 1368675_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400006749.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006760.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006748.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006756.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006759.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006754.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006757.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006751.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006752.rn.1 TC0400000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006755.rn.1 TC0400000947.rn.1 RTA TC
          Rat Expression Set 230 1368675_at - -
          Rat Expression Set 230 1368675_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855615 - -
          Rat Genome U34 Set rc_AI232194_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21603706 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376494 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline