Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00587905_m1
          See other CAP2 GE Assays ›
          Gene Symbol:
          Cap2
          Gene Name:
          CAP, adenylate cyclase-associated protein, 2 (yeast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 18441614 - 18592866 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00587905_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053874.1 NP_446326.1 3 - 4 273 64
          XM_006253753.2 XP_006253815.1 4 - 5 382 64
          XM_006253754.2 XP_006253816.1 4 - 5 495 64
          GenBank mRNA U31935.1 - 3 - 4 273 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001487.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cap2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          cytoskeleton organization
          actin cytoskeleton organization
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12967 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12967 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12967 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12967 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12967 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702155356 - -
          Agilent Rat cDNA Microarray (G4105A) 702155356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940555 - -
          Affymetrix Rat Gene 1.1 ST Array 10940555 - -
          Rat Genome 230 2.0 Array 1377788_PM_at - -
          Rat Genome 230 2.0 Array 1377788_at - -
          Rat Genome 230 2.0 Array 1368894_PM_at - -
          Rat Genome 230 2.0 Array 1368894_at - -
          Rat Genome 230 2.0 Array 1368893_PM_at - -
          Rat Genome 230 2.0 Array 1368893_at - -
          Affymetrix HT Rat Focus Array 1368893_PM_at - -
          Affymetrix HT Rat Focus Array 1368894_PM_at - -
          Affymetrix HT Rat Focus Array 1368893_at - -
          Affymetrix HT Rat Focus Array 1368894_at - -
          Affymetrix Rat Clariom S Assay PSR1700010407.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010396.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010399.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010403.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010400.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010397.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010398.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010404.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010401.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010406.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010411.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010405.rn.1 TC1700001487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700010402.rn.1 TC1700001487.rn.1 RTA TC
          Rat Expression Set 230 1368893_at - -
          Rat Expression Set 230 1368893_PM_at - -
          Rat Expression Set 230 1368894_at - -
          Rat Expression Set 230 1368894_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940555 - -
          Rat Genome U34 Set rc_AI145367_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22196553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline