Search
Search
| Gene Symbol: Otos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139188.3 | NP_631927.1 | 3 - 4 | 140 | 138 | |
| XM_008767358.1 | XP_008765580.1 | 4 - 5 | 748 | 138 | ||
| GenBank mRNA | AY062254.1 | - | 3 - 4 | 168 | 138 | |
Set Membership: |
|