Search
Search
| Gene Symbol: Lphn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130822.1 | NP_570835.1 | 16 - 17 | 3111 | 76 | |
| XM_008769968.1 | XP_008768190.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769969.1 | XP_008768191.1 | 16 - 17 | 3200 | 76 | ||
| XM_008769970.1 | XP_008768192.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769971.1 | XP_008768193.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769972.1 | XP_008768194.1 | 15 - 16 | 3176 | 76 | ||
| XM_008769973.1 | XP_008768195.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769974.1 | XP_008768196.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769975.1 | XP_008768197.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769976.1 | XP_008768198.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769977.1 | XP_008768199.1 | 15 - 16 | 3011 | 76 | ||
| XM_008769978.1 | XP_008768200.1 | 15 - 16 | 3176 | 76 | ||
| XM_008769979.1 | XP_008768201.1 | 15 - 16 | 3011 | 76 | ||
| XM_008769980.1 | XP_008768202.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769981.1 | XP_008768203.1 | 16 - 17 | 3215 | 76 | ||
| XM_008769982.1 | XP_008768204.1 | 16 - 17 | 3214 | 76 | ||
| XM_008769983.1 | XP_008768205.1 | 16 - 17 | 3214 | 76 | ||
| XM_008769984.1 | XP_008768206.1 | 16 - 17 | 3214 | 76 | ||
| XM_008769985.1 | XP_008768207.1 | 15 - 16 | 3010 | 76 | ||
| XM_008769986.1 | XP_008768208.1 | 15 - 16 | 3010 | 76 | ||
| GenBank mRNA | AF063103.1 | - | 16 - 17 | 3111 | 76 | |
| AF081154.1 | - | 15 - 16 | 2932 | 76 | ||
| AF081155.1 | - | 15 - 16 | 2932 | 76 | ||
| AF081156.1 | - | 15 - 16 | 2932 | 76 | ||
| AF081157.1 | - | 16 - 17 | 3136 | 76 | ||
| AF081158.1 | - | 16 - 17 | 3136 | 76 | ||
| AF081159.1 | - | 16 - 17 | 3136 | 76 | ||
Set Membership: |
|