Search
Search
| Gene Symbol: Ghrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012850.1 | NP_036982.1 | 9 - 10 | 911 | 93 | |
| XM_008762872.1 | XP_008761094.1 | 9 - 10 | 2274 | 93 | ||
| GenBank mRNA | AF122055.1 | - | 9 - 10 | 884 | 93 | |
| L01407.1 | - | 9 - 10 | 911 | 93 | ||
Set Membership: |
|