Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01334597_m1
          See other CTNND1 GE Assays ›
          Gene Symbol:
          Ctnnd1
          Gene Name:
          catenin (cadherin associated protein), delta 1
          Gene Aliases:
          Catns
          Chromosome Location:
          Chr.3: 72001904 - 72053044 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01334597_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctnnd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107740.1 NP_001101210.1 16 - 17 3047 97
          XM_006234452.2 XP_006234514.1 18 - 19 3236 97
          XM_006234453.2 XP_006234515.1 17 - 18 3118 97
          XM_006234454.2 XP_006234516.1 18 - 19 3236 97
          XM_006234456.2 XP_006234518.1 17 - 18 3218 97
          XM_006234459.2 XP_006234521.1 18 - 19 3236 97
          XM_006234460.2 XP_006234522.1 18 - 19 3236 97
          XM_008761987.1 XP_008760209.1 16 - 17 2829 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003432.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of a polarized epithelium
          cell adhesion
          brain development
          salivary gland morphogenesis
          single organismal cell-cell adhesion
          keratinocyte differentiation
          epithelial cell differentiation involved in salivary gland development
          negative regulation of canonical Wnt signaling pathway
          receptor binding
          protein binding
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          cadherin binding
          cell adhesion molecule binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P773732 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P297596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131048 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P297596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P773732 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P297596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P773732 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P297596 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P773732 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P297596 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P236532 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P837619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846810 - -
          Affymetrix Rat Gene 1.0 ST Array 10846809 - -
          Affymetrix Rat Gene 1.0 ST Array 10846816 - -
          Affymetrix Rat Gene 1.0 ST Array 10846803 - -
          Affymetrix Rat Gene 1.0 ST Array 10846811 - -
          Affymetrix Rat Gene 1.0 ST Array 10846812 - -
          Affymetrix Rat Gene 1.0 ST Array 10846805 - -
          Affymetrix Rat Gene 1.0 ST Array 10846799 - -
          Affymetrix Rat Gene 1.0 ST Array 10846804 - -
          Affymetrix Rat Gene 1.0 ST Array 10846798 - -
          Affymetrix Rat Gene 1.0 ST Array 10846813 - -
          Affymetrix Rat Gene 1.0 ST Array 10846795 - -
          Affymetrix Rat Gene 1.0 ST Array 10846801 - -
          Affymetrix Rat Gene 1.0 ST Array 10846806 - -
          Affymetrix Rat Gene 1.0 ST Array 10846797 - -
          Affymetrix Rat Gene 1.0 ST Array 10846807 - -
          Affymetrix Rat Gene 1.0 ST Array 10846802 - -
          Affymetrix Rat Gene 1.0 ST Array 10846808 - -
          Affymetrix Rat Gene 1.0 ST Array 10846814 - -
          Affymetrix Rat Gene 1.1 ST Array 10846794 - -
          Rat Genome 230 2.0 Array 1388415_at - -
          Rat Genome 230 2.0 Array 1388415_PM_at - -
          Affymetrix HT Rat Focus Array 1388415_at - -
          Affymetrix HT Rat Focus Array 1388415_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300026991.rn.1 TC0300003432.rn.1 RTA TC
          Rat Expression Set 230 1388415_PM_at - -
          Rat Expression Set 230 1388415_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846794 - -
          Rat Genome U34 Set rc_AI012246_at - -
          Rat Genome U34 Set rc_AI105019_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline