Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01400464_m1
See other SERPINE2 GE Assays ›
Gene Symbol:
Serpine2
Gene Name:
serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2
Gene Aliases:
Gdnpn1, PI-7, Pn-1, Spin4
Chromosome Location:
Chr.9: 85560852 - 85626125 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01400464_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Serpine2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_019197.1
NP_062070.1
1 - 2
103
86
GenBank mRNA
CK357674.1
-
1 - 2
103
86
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_019197.1 transcripts.
Set Membership:
Amplicon length between 86 and 100
Rattus norvegicus
Probe spans exons
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
embryo implantation
blood coagulation
negative regulation of cell proliferation
response to radiation
response to wounding
negative regulation of peptidase activity
negative regulation of platelet activation
negative regulation of plasminogen activation
negative regulation of sodium ion transport
negative regulation of endopeptidase activity
positive regulation of neuron projection development
negative regulation of phosphatidylinositol 3-kinase signaling
cerebellar granular layer morphogenesis
negative regulation of blood coagulation
negative regulation of cell growth
secretion by cell
secretory granule organization
negative regulation of protein catabolic process
mating plug formation
negative regulation of proteolysis
negative regulation of smoothened signaling pathway
regulation of timing of cell differentiation
positive regulation of astrocyte differentiation
detection of mechanical stimulus involved in sensory perception
regulation of synaptic transmission, glutamatergic
long-term synaptic potentiation
innervation
seminal vesicle epithelium development
dense core granule biogenesis
negative regulation of platelet aggregation
serine-type endopeptidase inhibitor activity
receptor binding
glycosaminoglycan binding
heparin binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0900018984.rn.1
TC0900002315.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0900018980.rn.1
TC0900002315.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline