Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01410595_m1
          See other CCDC172 GE Assays ›
          Gene Symbol:
          Ccdc172
          Gene Name:
          coiled-coil domain containing 172
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 279633655 - 279682008 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01410595_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc172
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014169.2 NP_001014191.1 5 - 6 539 89
          XM_006231674.2 XP_006231736.1 5 - 6 555 89
          XM_008760539.1 XP_008758761.1 4 - 5 828 89
          XM_008760540.1 XP_008758762.1 3 - 4 711 89
          XM_008760541.1 XP_008758763.1 4 - 5 828 89
          XM_008760542.1 XP_008758764.1 4 - 5 828 89
          GenBank mRNA BC079324.1 - 4 - 5 756 7135457 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc172

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92959 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36607 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011969 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28953 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716371 - -
          Affymetrix Rat Gene 1.0 ST Array 10716372 - -
          Affymetrix Rat Gene 1.0 ST Array 10716368 - -
          Affymetrix Rat Gene 1.0 ST Array 10716370 - -
          Affymetrix Rat Gene 1.0 ST Array 10716369 - -
          Affymetrix Rat Gene 1.0 ST Array 10716367 - -
          Affymetrix Rat Gene 1.0 ST Array 10716366 - -
          Affymetrix Rat Gene 1.1 ST Array 10716365 - -
          Affymetrix Rat Clariom S Assay PSR0100031770.rn.1 TC0100003886.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10716365 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21889713 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650147 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline