Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412143_m1
          See other CADM3 GE Assays ›
          Gene Symbol:
          Cadm3
          Gene Name:
          cell adhesion molecule 3
          Gene Aliases:
          Igsf4b, Necl-1
          Chromosome Location:
          Chr.13: 91842311 - 91873123 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412143_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cadm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047103.1 NP_001040568.1 8 - 9 1072 109
          XM_006250323.2 XP_006250385.1 9 - 10 1344 109
          GenBank mRNA BC161811.1 - 8 - 9 1073 7317639 109
          DQ272743.1 - 8 - 9 1072 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001972.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          protein localization
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P454645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P454645 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P454645 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P454645 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94051 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76149 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9109 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770077 - -
          Affymetrix Rat Gene 1.0 ST Array 10770078 - -
          Affymetrix Rat Gene 1.0 ST Array 10770076 - -
          Affymetrix Rat Gene 1.0 ST Array 10770072 - -
          Affymetrix Rat Gene 1.0 ST Array 10770073 - -
          Affymetrix Rat Gene 1.0 ST Array 10770079 - -
          Affymetrix Rat Gene 1.0 ST Array 10770074 - -
          Affymetrix Rat Gene 1.0 ST Array 10770075 - -
          Affymetrix Rat Gene 1.0 ST Array 10770071 - -
          Affymetrix Rat Gene 1.1 ST Array 10770070 - -
          Rat Genome 230 2.0 Array 1390033_PM_at - -
          Rat Genome 230 2.0 Array 1394668_at - -
          Rat Genome 230 2.0 Array 1390033_at - -
          Rat Genome 230 2.0 Array 1394668_PM_at - -
          Affymetrix HT Rat Focus Array 1394668_at - -
          Affymetrix HT Rat Focus Array 1394668_PM_at - -
          Affymetrix HT Rat Focus Array 1390033_at - -
          Affymetrix HT Rat Focus Array 1390033_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300014159.rn.1 TC1300001972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014158.rn.1 TC1300001972.rn.1 RTA TC
          Rat Expression Set 230 1390033_PM_at - -
          Rat Expression Set 230 1390033_at - -
          Rat Expression Set 230 1394668_PM_at - -
          Rat Expression Set 230 1394668_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770070 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359083 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline