Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01412640_m1
          See other ZGPAT GE Assays ›
          Gene Symbol:
          Zgpat
          Gene Name:
          zinc finger, CCCH-type with G patch domain
          Gene Aliases:
          RGD1310801
          Chromosome Location:
          Chr.3: 176865113 - 176881603 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01412640_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zgpat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009656.1 NP_001009656.1 3 - 4 993 94
          XM_006235749.1 XP_006235811.1 4 - 5 989 94
          XM_006235750.1 XP_006235812.1 4 - 5 954 94
          XM_006235751.2 XP_006235813.1 4 - 5 975 94
          XM_006235752.2 XP_006235814.1 3 - 4 889 94
          GenBank mRNA BC087720.1 - 3 - 4 993 7318724 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002599.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zgpat

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          negative regulation of epidermal growth factor-activated receptor activity
          negative regulation of transcription, DNA-templated
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28216 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133139 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133139 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133139 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P702623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70713 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75428 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843020 - -
          Affymetrix Rat Gene 1.0 ST Array 10843018 - -
          Affymetrix Rat Gene 1.0 ST Array 10843016 - -
          Affymetrix Rat Gene 1.0 ST Array 10843014 - -
          Affymetrix Rat Gene 1.0 ST Array 10843015 - -
          Affymetrix Rat Gene 1.0 ST Array 10843019 - -
          Affymetrix Rat Gene 1.1 ST Array 10843013 - -
          Rat Genome 230 2.0 Array 1374147_PM_at - -
          Rat Genome 230 2.0 Array 1374147_at - -
          Rat Genome 230 2.0 Array 1378319_PM_at - -
          Rat Genome 230 2.0 Array 1391044_s_at - -
          Rat Genome 230 2.0 Array 1378319_at - -
          Rat Genome 230 2.0 Array 1391044_PM_s_at - -
          Affymetrix HT Rat Focus Array 1374147_at - -
          Affymetrix HT Rat Focus Array 1374147_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300019231.rn.1 TC0300002599.rn.1 RTA TC
          Rat Expression Set 230 1374147_PM_at - -
          Rat Expression Set 230 1374147_at - -
          Rat Expression Set 230 1391044_s_at - -
          Rat Expression Set 230 1391044_PM_s_at - -
          Rat Expression Set 230 1378319_PM_at - -
          Rat Expression Set 230 1378319_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843013 - -
          Rat Genome U34 Set rc_AA924186_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21335724 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363578 - -
          RatRef-12 Expression BeadChip ILMN_53093 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline