Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424029_m1
          See other SGK2 GE Assays ›
          Gene Symbol:
          Sgk2
          Gene Name:
          serum/glucocorticoid regulated kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 159361313 - 159385843 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424029_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sgk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134463.1 NP_604458.1 6 - 7 767 72
          GenBank mRNA AF361756.1 - 3 - 4 180 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002248.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein serine/threonine kinase activity
          ATP binding
          potassium channel regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P477827 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103911 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14040 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14040 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P477827 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14040 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P477827 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2601 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841957 - -
          Affymetrix Rat Gene 1.0 ST Array 10841948 - -
          Affymetrix Rat Gene 1.0 ST Array 10841960 - -
          Affymetrix Rat Gene 1.0 ST Array 10841959 - -
          Affymetrix Rat Gene 1.0 ST Array 10841954 - -
          Affymetrix Rat Gene 1.0 ST Array 10841951 - -
          Affymetrix Rat Gene 1.0 ST Array 10841956 - -
          Affymetrix Rat Gene 1.0 ST Array 10841953 - -
          Affymetrix Rat Gene 1.0 ST Array 10841952 - -
          Affymetrix Rat Gene 1.0 ST Array 10841950 - -
          Affymetrix Rat Gene 1.0 ST Array 10841958 - -
          Affymetrix Rat Gene 1.0 ST Array 10841955 - -
          Affymetrix Rat Gene 1.1 ST Array 10841946 - -
          Rat Genome 230 2.0 Array 1380262_at - -
          Rat Genome 230 2.0 Array 1380262_PM_at - -
          Affymetrix HT Rat Focus Array 1380262_at - -
          Affymetrix HT Rat Focus Array 1380262_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300016736.rn.1 TC0300002248.rn.1 RTA TC
          Rat Expression Set 230 1380262_PM_at - -
          Rat Expression Set 230 1380262_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841946 - -
          Rat Genome U34 Set rc_AA893436_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22416795 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353015 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline