Search
Search
| Gene Symbol: Rev1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108213.1 | NP_001101683.1 | 8 - 9 | 1550 | 153 | |
| XM_006244819.2 | XP_006244881.1 | 9 - 10 | 2282 | 153 | ||
| XM_006244820.2 | XP_006244882.1 | 9 - 10 | 2902 | 153 | ||
| XM_006244821.2 | XP_006244883.1 | 8 - 9 | 2081 | 153 | ||
| XM_008767030.1 | XP_008765252.1 | 5 - 6 | 1089 | 153 | ||
Set Membership: |
|