Search
Search
| Gene Symbol: Asmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144759.2 | NP_653360.2 | 1 - 2 | 100 | 51 | |
| XM_006248991.2 | XP_006249053.1 | 1 - 2 | 886 | 51 | ||
| GenBank mRNA | CB765131.1 | - | 1 - 2 | 100 | 51 | |
| EU741665.1 | - | 1 - 2 | 80 | 51 | ||
Set Membership: |
|