Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454643_m1
          See other LIPK GE Assays ›
          Gene Symbol:
          Lipk
          Gene Name:
          lipase, family member K
          Gene Aliases:
          Lipl2
          Chromosome Location:
          Chr.1: 252323865 - 252355541 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454643_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106374.1 NP_001099844.1 9 - 10 1060 77
          XM_008760325.1 XP_008758547.1 10 - 11 1068 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003506.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          hydrolase lipase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid catabolic process
          hydrolase activity, acting on ester bonds

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167762 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002576 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231769 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714860 - -
          Affymetrix Rat Gene 1.0 ST Array 10714861 - -
          Affymetrix Rat Gene 1.0 ST Array 10714864 - -
          Affymetrix Rat Gene 1.0 ST Array 10714863 - -
          Affymetrix Rat Gene 1.0 ST Array 10714862 - -
          Affymetrix Rat Gene 1.0 ST Array 10714859 - -
          Affymetrix Rat Gene 1.0 ST Array 10714865 - -
          Affymetrix Rat Gene 1.0 ST Array 10714867 - -
          Affymetrix Rat Gene 1.0 ST Array 10714866 - -
          Affymetrix Rat Gene 1.0 ST Array 10714868 - -
          Affymetrix Rat Gene 1.1 ST Array 10714858 - -
          Affymetrix Rat Clariom S Assay PSR0100028409.rn.1 TC0100003506.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028410.rn.1 TC0100003506.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10714858 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22322040 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368254 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline