Search
Search
| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031730.2 | NP_113918.2 | 3 - 4 | 1926 | 60 | |
| XM_008762830.1 | XP_008761052.1 | 3 - 4 | 450 | 60 | ||
| GenBank mRNA | CO393575.1 | - | 3 - 4 | 487 | 60 | |
| M59980.1 | - | 3 - 4 | 1605 | 60 | ||
| S64320.1 | - | 3 - 4 | 1924 | 60 | ||
Set Membership: |
|