Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458715_g1
          See other SHC1 GE Assays ›
          Gene Symbol:
          Shc1
          Gene Name:
          SHC (Src homology 2 domain containing) transforming protein 1
          Gene Aliases:
          P66shc
          Chromosome Location:
          Chr.2: 188745503 - 188757066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01458715_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Shc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164060.1 NP_001157532.1 8 - 9 1377 87
          NM_053517.2 NP_445969.2 9 - 10 944 87
          GenBank mRNA BC088298.1 - 9 - 10 944 7323045 87
          CB813924.1 - 2 - 3 224 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Shc1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          angiogenesis
          response to hypoxia
          epidermal growth factor receptor signaling pathway
          heart development
          aging
          insulin receptor signaling pathway
          response to toxic substance
          response to organic cyclic compound
          single organismal cell-cell adhesion
          actin cytoskeleton organization
          neuron differentiation
          organ regeneration
          neuron projection development
          actin cytoskeleton reorganization
          response to insulin
          cellular response to insulin stimulus
          response to nicotine
          intracellular signal transduction
          regulation of growth
          regulation of cell proliferation
          response to hydrogen peroxide
          positive regulation of DNA replication
          positive regulation of vasoconstriction
          positive regulation of smooth muscle cell proliferation
          response to glucocorticoid
          phosphotyrosine binding
          epidermal growth factor receptor binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          neurotrophin TRKA receptor binding
          protein binding
          protein kinase binding
          receptor tyrosine kinase binding
          protein complex binding
          ephrin receptor binding
          phosphoprotein binding
          protein phosphatase 2A binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P106779 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88970 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019295 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019295 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106779 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019295 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P106779 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P445306 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816798 - -
          Affymetrix Rat Gene 1.0 ST Array 10816800 - -
          Affymetrix Rat Gene 1.0 ST Array 10816796 - -
          Affymetrix Rat Gene 1.0 ST Array 10816794 - -
          Affymetrix Rat Gene 1.0 ST Array 10816795 - -
          Affymetrix Rat Gene 1.0 ST Array 10816799 - -
          Affymetrix Rat Gene 1.0 ST Array 10816797 - -
          Affymetrix Rat Gene 1.0 ST Array 10816792 - -
          Affymetrix Rat Gene 1.1 ST Array 10816791 - -
          Rat Genome 230 2.0 Array 1377594_PM_at - -
          Rat Genome 230 2.0 Array 1377594_at - -
          Affymetrix HT Rat Focus Array 1377594_PM_at - -
          Affymetrix HT Rat Focus Array 1377594_at - -
          Affymetrix Rat Clariom S Assay PSR0200034713.rn.1 TC0200004978.rn.1 RTA TC
          Rat Expression Set 230 1377594_at - -
          Rat Expression Set 230 1377594_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816791 - -
          Rat Genome U34 Set rc_AA899410_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20910974 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356122 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline