Search
Search

Slco1a6| Gene Symbol: Slco1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130736.1 | NP_570092.1 | 6 - 7 | 721 | 109 | |
| XM_006237650.2 | XP_006237712.1 | 7 - 8 | 775 | 109 | ||
| GenBank mRNA | AF053317.2 | - | 6 - 7 | 721 | 109 | |
Set Membership: |
|