Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477735_m1
          See other RGMA GE Assays ›
          Gene Symbol:
          Rgma
          Gene Name:
          repulsive guidance molecule family member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 134699053 - 134742512 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgma
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107524.1 NP_001100994.1 3 - 4 999 123
          XM_008759521.1 XP_008757743.1 1 - 2 2446 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001764.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          BMP signaling pathway
          regulation of BMP signaling pathway
          coreceptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117510 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10083 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10083 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117510 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P485067 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707936 - -
          Affymetrix Rat Gene 1.0 ST Array 10707938 - -
          Affymetrix Rat Gene 1.0 ST Array 10707937 - -
          Affymetrix Rat Gene 1.1 ST Array 10707935 - -
          Rat Genome 230 2.0 Array 1375849_at - -
          Rat Genome 230 2.0 Array 1375849_PM_at - -
          Affymetrix HT Rat Focus Array 1375849_at - -
          Affymetrix HT Rat Focus Array 1375849_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100013994.rn.1 TC0100001764.rn.1 RTA TC
          Rat Expression Set 230 1375849_at - -
          Rat Expression Set 230 1375849_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10707935 - -
          Rat Genome U34 Set rc_AA819234_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21753687 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline