Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01480291_m1
          See other VPS33A GE Assays ›
          Gene Symbol:
          Vps33a
          Gene Name:
          vacuolar protein sorting 33 homolog A (S. cerevisiae)
          Gene Aliases:
          r-Vps33a
          Chromosome Location:
          Chr.12: 38459816 - 38482903 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01480291_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vps33a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022961.3 NP_075250.2 11 - 12 1633 159
          GenBank mRNA BC105752.1 - 11 - 12 1633 5599123 159

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vps33a

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle docking involved in exocytosis
          biological_process
          protein transport
          vesicle-mediated transport
          platelet formation
          melanosome localization
          lysosome localization
          pigmentation
          regulation of developmental pigmentation
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1060167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62095 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1060167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1060167 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1060167 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1060170 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1060167 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12354 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P318604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98920 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78684 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21424 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758420 - -
          Affymetrix Rat Gene 1.0 ST Array 10758419 - -
          Affymetrix Rat Gene 1.0 ST Array 10758426 - -
          Affymetrix Rat Gene 1.0 ST Array 10758418 - -
          Affymetrix Rat Gene 1.0 ST Array 10758428 - -
          Affymetrix Rat Gene 1.0 ST Array 10758429 - -
          Affymetrix Rat Gene 1.0 ST Array 10758423 - -
          Affymetrix Rat Gene 1.0 ST Array 10758421 - -
          Affymetrix Rat Gene 1.0 ST Array 10758427 - -
          Affymetrix Rat Gene 1.0 ST Array 10758422 - -
          Affymetrix Rat Gene 1.0 ST Array 10758424 - -
          Affymetrix Rat Gene 1.0 ST Array 10758417 - -
          Affymetrix Rat Gene 1.0 ST Array 10758425 - -
          Affymetrix Rat Gene 1.0 ST Array 10758416 - -
          Affymetrix Rat Gene 1.1 ST Array 10758415 - -
          Rat Genome 230 2.0 Array 1382156_at - -
          Rat Genome 230 2.0 Array 1385081_PM_s_at - -
          Rat Genome 230 2.0 Array 1382157_s_at - -
          Rat Genome 230 2.0 Array 1385081_s_at - -
          Rat Genome 230 2.0 Array 1387557_s_at - -
          Rat Genome 230 2.0 Array 1387557_PM_s_at - -
          Rat Genome 230 2.0 Array 1382157_PM_s_at - -
          Rat Genome 230 2.0 Array 1382156_PM_at - -
          Affymetrix HT Rat Focus Array 1387557_PM_s_at - -
          Affymetrix HT Rat Focus Array 1385081_s_at - -
          Affymetrix HT Rat Focus Array 1385081_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387557_s_at - -
          Rat Expression Set 230 1387557_PM_s_at - -
          Rat Expression Set 230 1387557_s_at - -
          Rat Expression Set 230 1385081_PM_s_at - -
          Rat Expression Set 230 1385081_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758415 - -
          Rat Genome U34 Set rc_AA900601_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366105 - -
          RatRef-12 Expression BeadChip ILMN_54492 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline