Search
Search
| Gene Symbol: Gpx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106411.1 | NP_001099881.1 | 2 - 3 | 502 | 125 | |
| GenBank mRNA | FQ214900.1 | - | 2 - 3 | 476 | 125 | |
| FQ220116.1 | - | 2 - 3 | 473 | 125 | ||
| FQ220265.1 | - | 2 - 3 | 482 | 125 | ||
| FQ220543.1 | - | 2 - 3 | 482 | 125 | ||
| FQ229597.1 | - | 2 - 3 | 476 | 125 | ||
| FQ229607.1 | - | 2 - 3 | 472 | 125 | ||
| FQ229635.1 | - | 2 - 3 | 476 | 125 | ||
| FQ229721.1 | - | 2 - 3 | 476 | 125 | ||
| FQ230033.1 | - | 2 - 3 | 476 | 125 | ||
Set Membership: |
|