Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497228_m1
          See other CENPI GE Assays ›
          Gene Symbol:
          Cenpi
          Gene Name:
          centromere protein I
          Gene Aliases:
          Fshprh1, LRPR1, RNALRP
          Chromosome Location:
          Chr.X: 105147061 - 105198798 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497228_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cenpi
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012955.1 NP_037087.1 21 - 22 2323 104
          XM_006257215.2 XP_006257277.1 21 - 22 2461 104
          XM_006257216.2 XP_006257278.1 21 - 22 2622 104
          XM_006257217.2 XP_006257279.1 20 - 21 2336 104
          XM_006257218.1 XP_006257280.1 19 - 20 1988 104
          XM_008773403.1 XP_008771625.1 20 - 21 2677 104
          GenBank mRNA X90355.1 - 21 - 22 2323 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000874.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cenpi

          Gene Ontology Categories:

          Function(s)

          centromere complex assembly
          follicle-stimulating hormone signaling pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P473159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70108 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P473159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P473159 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P473159 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78438 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104734 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934909 - -
          Affymetrix Rat Gene 1.0 ST Array 10934902 - -
          Affymetrix Rat Gene 1.0 ST Array 10934905 - -
          Affymetrix Rat Gene 1.0 ST Array 10934896 - -
          Affymetrix Rat Gene 1.0 ST Array 10934894 - -
          Affymetrix Rat Gene 1.0 ST Array 10934899 - -
          Affymetrix Rat Gene 1.0 ST Array 10934904 - -
          Affymetrix Rat Gene 1.0 ST Array 10934912 - -
          Affymetrix Rat Gene 1.0 ST Array 10934892 - -
          Affymetrix Rat Gene 1.0 ST Array 10934913 - -
          Affymetrix Rat Gene 1.0 ST Array 10934901 - -
          Affymetrix Rat Gene 1.0 ST Array 10934895 - -
          Affymetrix Rat Gene 1.0 ST Array 10934910 - -
          Affymetrix Rat Gene 1.0 ST Array 10934903 - -
          Affymetrix Rat Gene 1.0 ST Array 10934907 - -
          Affymetrix Rat Gene 1.0 ST Array 10934906 - -
          Affymetrix Rat Gene 1.0 ST Array 10934908 - -
          Affymetrix Rat Gene 1.0 ST Array 10934900 - -
          Affymetrix Rat Gene 1.0 ST Array 10934898 - -
          Affymetrix Rat Gene 1.0 ST Array 10934911 - -
          Affymetrix Rat Gene 1.0 ST Array 10934893 - -
          Affymetrix Rat Gene 1.0 ST Array 10934897 - -
          Affymetrix Rat Gene 1.1 ST Array 10934891 - -
          Rat Genome 230 2.0 Array 1368773_at - -
          Rat Genome 230 2.0 Array 1368773_PM_at - -
          Affymetrix HT Rat Focus Array 1368773_PM_at - -
          Affymetrix HT Rat Focus Array 1368773_at - -
          Affymetrix Rat Clariom S Assay PSR0X00005181.rn.1 TC0X00000874.rn.1 RTA TC
          Rat Expression Set 230 1368773_PM_at - -
          Rat Expression Set 230 1368773_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934891 - -
          Rat Genome U34 Set X90355_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21572978 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371854 - -
          RatRef-12 Expression BeadChip ILMN_70247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline