Search
Search
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241616.1 | XP_006241678.1 | 2 - 3 | 180 | 129 | |
| XM_006241617.1 | XP_006241679.1 | 2 - 3 | 180 | 129 | ||
| GenBank mRNA | FQ211706.1 | - | 2 - 3 | 180 | 129 | |
Set Membership: |
|