Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504470_m1
          See other RIMS2 GE Assays ›
          Gene Symbol:
          Rims2
          Gene Name:
          regulating synaptic membrane exocytosis 2
          Gene Aliases:
          Nim2, Rim2
          Chromosome Location:
          Chr.7: 78092020 - 78594162 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504470_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rims2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053945.2 NP_446397.1 8 - 9 2087 151
          XM_006241591.2 XP_006241653.1 8 - 9 2245 151
          XM_006241592.2 XP_006241654.1 8 - 9 2245 151
          XM_006241593.2 XP_006241655.1 8 - 9 2103 151
          XM_006241594.2 XP_006241656.1 8 - 9 2103 151
          XM_006241598.2 XP_006241660.1 6 - 7 4859 151
          XM_008765437.1 XP_008763659.1 10 - 11 4399 151
          XM_008765438.1 XP_008763660.1 8 - 9 2245 151
          XM_008765439.1 XP_008763661.1 9 - 10 4387 151
          XM_008765440.1 XP_008763662.1 8 - 9 2245 151
          XM_008765441.1 XP_008763663.1 8 - 9 2104 151
          XM_008765442.1 XP_008763664.1 8 - 9 2104 151
          XM_008765443.1 XP_008763665.1 8 - 9 2103 151
          XM_008765444.1 XP_008763666.1 10 - 11 4401 151
          XM_008765447.1 XP_008763669.1 10 - 11 2365 151
          GenBank mRNA AF199322.1 - 8 - 9 2087 151
          AF199323.1 - 8 - 9 2087 151
          AF199324.1 - 8 - 9 2228 151
          AF199325.1 - 8 - 9 2087 151
          AF199326.1 - 8 - 9 2087 151
          AF199327.1 - 8 - 9 2087 151
          AF199328.1 - 8 - 9 2087 151
          AF199329.1 - 8 - 9 2087 151
          AF199330.1 - 8 - 9 2087 151
          AF199331.1 - 8 - 9 2087 151
          AF199332.1 - 8 - 9 2087 151
          AF548738.1 - 6 - 7 1510 151

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001018.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rims2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          exocytosis
          positive regulation of gene expression
          calcium ion regulated exocytosis
          regulation of exocytosis
          cAMP-mediated signaling
          insulin secretion
          regulation of membrane potential
          calcium ion-regulated exocytosis of neurotransmitter
          positive regulation of inhibitory postsynaptic potential
          positive regulation of excitatory postsynaptic potential
          protein binding
          Rab GTPase binding
          protein domain specific binding
          ion channel binding
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0700007681.rn.1 TC0700001018.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline