Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01512149_m1
          See other MRPL14 GE Assays ›
          Gene Symbol:
          Mrpl14
          Gene Name:
          mitochondrial ribosomal protein L14
          Gene Aliases:
          L14mt, L32mt, MRP-L14, MRP-L32
          Chromosome Location:
          Chr.9: 17687622 - 17698588 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01512149_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106890.2 NP_001100360.1 2 - 3 210 91
          NM_001271131.1 NP_001258060.1 2 - 3 166 91
          XM_006244512.2 XP_006244574.1 2 - 3 400 91
          XM_008766849.1 XP_008765071.1 3 - 4 333 91
          XM_008766850.1 XP_008765072.1 2 - 3 223 91
          GenBank mRNA FM100935.1 - 2 - 3 140 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001602.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045957 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045957 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P133611 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926578 - -
          Affymetrix Rat Gene 1.0 ST Array 10926580 - -
          Affymetrix Rat Gene 1.0 ST Array 10926579 - -
          Affymetrix Rat Gene 1.1 ST Array 10926577 - -
          Rat Genome 230 2.0 Array 1395063_at - -
          Rat Genome 230 2.0 Array 1395063_PM_at - -
          Rat Genome 230 2.0 Array 1372597_PM_at - -
          Rat Genome 230 2.0 Array 1372597_at - -
          Affymetrix HT Rat Focus Array 1372597_PM_at - -
          Affymetrix HT Rat Focus Array 1372597_at - -
          Affymetrix Rat Clariom S Assay PSR0900013359.rn.1 TC0900001602.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013363.rn.1 TC0900001602.rn.1 RTA TC
          Rat Expression Set 230 1372597_PM_at - -
          Rat Expression Set 230 1372597_at - -
          Rat Expression Set 230 1395063_at - -
          Rat Expression Set 230 1395063_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926577 - -
          Rat Genome U34 Set rc_AA900976_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21337710 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline