Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518771_m1
          See other ZDHHC18 GE Assays ›
          Gene Symbol:
          Zdhhc18
          Gene Name:
          zinc finger, DHHC-type containing 18
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 151829795 - 151856256 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518771_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zdhhc18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039339.1 NP_001034428.1 7 - 8 1078 81
          GenBank mRNA AY886534.1 - 7 - 8 1078 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003781.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          protein palmitoylation
          cellular protein localization
          zinc ion binding
          palmitoyltransferase activity
          protein-cysteine S-palmitoyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036761 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P915168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210514 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P915168 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P915168 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036761 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P915168 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P915168 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P736967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872566 - -
          Affymetrix Rat Gene 1.0 ST Array 10872568 - -
          Affymetrix Rat Gene 1.0 ST Array 10872569 - -
          Affymetrix Rat Gene 1.0 ST Array 10872565 - -
          Affymetrix Rat Gene 1.0 ST Array 10872571 - -
          Affymetrix Rat Gene 1.0 ST Array 10872563 - -
          Affymetrix Rat Gene 1.0 ST Array 10872564 - -
          Affymetrix Rat Gene 1.0 ST Array 10872570 - -
          Affymetrix Rat Gene 1.1 ST Array 10872561 - -
          Rat Genome 230 2.0 Array 1375096_PM_at - -
          Rat Genome 230 2.0 Array 1375096_at - -
          Rat Genome 230 2.0 Array 1374306_at - -
          Rat Genome 230 2.0 Array 1374306_PM_at - -
          Affymetrix HT Rat Focus Array 1374306_at - -
          Affymetrix HT Rat Focus Array 1374306_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028272.rn.1 TC0500003781.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028273.rn.1 TC0500003781.rn.1 RTA TC
          Rat Expression Set 230 1374306_at - -
          Rat Expression Set 230 1374306_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872561 - -
          Rat Genome U34 Set rc_AI237254_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22169514 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355425 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline