Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01524182_g1
          See other TCP1 GE Assays ›
          Gene Symbol:
          Tcp1
          Gene Name:
          t-complex 1
          Gene Aliases:
          CCTalpha, Tcp-1
          Chromosome Location:
          Chr.1: 48025664 - 48033343 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01524182_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012670.1 NP_036802.1 10 - 11 1370 76
          GenBank mRNA D90345.1 - 10 - 11 1370 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcp1

          Panther Classification:

          Molecular Function -

          chaperonin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          spermatid development
          binding of sperm to zona pellucida
          translocation of peptides or proteins into host cell cytoplasm
          cellular protein metabolic process
          toxin transport
          regulation of macrophage apoptotic process
          ATP binding
          ubiquitin protein ligase binding
          poly(A) RNA binding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P675372 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71394 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P675372 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P675372 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035058 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P675372 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78096 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863889 - -
          Affymetrix Rat Gene 1.0 ST Array 10717946 - -
          Affymetrix Rat Gene 1.0 ST Array 10717943 - -
          Affymetrix Rat Gene 1.0 ST Array 10717942 - -
          Affymetrix Rat Gene 1.0 ST Array 10717944 - -
          Affymetrix Rat Gene 1.0 ST Array 10717950 - -
          Affymetrix Rat Gene 1.0 ST Array 10717945 - -
          Affymetrix Rat Gene 1.0 ST Array 10863887 - -
          Affymetrix Rat Gene 1.1 ST Array 10863886 - -
          Affymetrix Rat Gene 1.1 ST Array 10717941 - -
          Rat Genome 230 2.0 Array 1368049_PM_at - -
          Rat Genome 230 2.0 Array 1368049_at - -
          Affymetrix HT Rat Focus Array 1368049_at - -
          Affymetrix HT Rat Focus Array 1368049_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100035345.rn.1 TC0100004441.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100035346.rn.1 TC0100004441.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100035347.rn.1 TC0100004441.rn.1 RTA TC
          Rat Expression Set 230 1368049_at - -
          Rat Expression Set 230 1368049_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10717941 - -
          Affymetrix Rat Gene 1.0 ST Array 10717949 - -
          Affymetrix Rat Gene 1.0 ST Array 10863888 - -
          Rat Genome U34 Set rc_AA900850_at - -
          Rat Genome U34 Set rc_AA900850_g_at - -
          Rat Genome U34 Set rc_AA875054_at - -
          Rat Genome U34 Set rc_AA875054_at - -
          Rat Genome U34 Set rc_AA901217_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22044554 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360156 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359719 - -
          RatRef-12 Expression BeadChip ILMN_59797 - -
          RatRef-12 Expression BeadChip ILMN_51744 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline