Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01527891_m1
See other HSD17B3 GE Assays ›
Gene Symbol:
Hsd17b3
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 1579319 - 1610745 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01527891_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Hsd17b3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_054007.1
NP_446459.1
5 - 6
461
82
GenBank mRNA
AF035156.1
-
5 - 6
461
82
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1700001181.rn.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Rattus norvegicus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Hsd17b3
Gene Ontology Categories:
Function(s)
Process(es)
organic acid metabolic process
steroid biosynthetic process
response to herbicide
response to metal ion
response to lead ion
response to organic cyclic compound
response to activity
response to insecticide
insecticide metabolic process
biphenyl metabolic process
phenol-containing compound metabolic process
hippocampus development
response to nutrient levels
cellular response to hormone stimulus
response to vitamin E
Leydig cell differentiation
response to L-ascorbic acid
response to estrogen
response to ethanol
response to arsenic-containing substance
response to cadmium ion
response to steroid hormone
response to corticosterone
oxidation-reduction process
response to fungicide
testosterone biosynthetic process
response to fatty acid
cellular response to antibiotic
cellular response to gonadotropin stimulus
estradiol 17-beta-dehydrogenase activity
oxidoreductase activity
testosterone dehydrogenase (NAD+) activity
testosterone 17-beta-dehydrogenase (NADP+) activity
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1700008118.rn.1
TC1700001181.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1700008116.rn.1
TC1700001181.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1700008117.rn.1
TC1700001181.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline