Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533350_m1
          See other RARG GE Assays ›
          Gene Symbol:
          Rarg
          Gene Name:
          retinoic acid receptor, gamma
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 143840739 - 143863206 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533350_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rarg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135249.1 NP_001128721.1 5 - 6 965 80
          NM_001135250.1 NP_001128722.1 3 - 4 737 80
          XM_008765784.1 XP_008764006.1 4 - 5 938 80
          XM_008765785.1 XP_008764007.1 5 - 6 929 80
          XM_008765786.1 XP_008764008.1 4 - 5 1457 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003968.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Nuclear Receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural tube closure
          chondrocyte development
          glandular epithelial cell development
          retinal pigment epithelium development
          growth plate cartilage development
          growth plate cartilage chondrocyte growth
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          anterior/posterior pattern specification
          regulation of gene expression
          positive regulation of gene expression
          embryonic camera-type eye development
          regulation of myelination
          negative regulation of chondrocyte differentiation
          response to retinoic acid
          embryonic hindlimb morphogenesis
          multicellular organism growth
          camera-type eye development
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          steroid hormone mediated signaling pathway
          negative regulation of cell differentiation
          regulation of myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          embryonic eye morphogenesis
          retinoic acid receptor signaling pathway
          reproductive structure development
          gland development
          retina development in camera-type eye
          limb development
          face development
          bone development
          bone morphogenesis
          epithelium development
          trachea cartilage development
          prostate gland epithelium morphogenesis
          negative regulation of cartilage development
          Harderian gland development
          cellular response to retinoic acid
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          retinoic acid receptor activity
          zinc ion binding
          sequence-specific DNA binding
          retinoid X receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119572 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46655 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119572 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16284 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119572 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907578 - -
          Affymetrix Rat Gene 1.0 ST Array 10907582 - -
          Affymetrix Rat Gene 1.0 ST Array 10907576 - -
          Affymetrix Rat Gene 1.0 ST Array 10907583 - -
          Affymetrix Rat Gene 1.0 ST Array 10907584 - -
          Affymetrix Rat Gene 1.0 ST Array 10907580 - -
          Affymetrix Rat Gene 1.0 ST Array 10907581 - -
          Affymetrix Rat Gene 1.0 ST Array 10907577 - -
          Affymetrix Rat Gene 1.0 ST Array 10907575 - -
          Affymetrix Rat Gene 1.0 ST Array 10907579 - -
          Affymetrix Rat Gene 1.1 ST Array 10907574 - -
          Rat Genome 230 2.0 Array 1376023_at - -
          Rat Genome 230 2.0 Array 1390771_PM_at - -
          Rat Genome 230 2.0 Array 1376023_PM_at - -
          Rat Genome 230 2.0 Array 1390771_at - -
          Affymetrix HT Rat Focus Array 1376023_at - -
          Affymetrix HT Rat Focus Array 1390771_at - -
          Affymetrix HT Rat Focus Array 1376023_PM_at - -
          Affymetrix HT Rat Focus Array 1390771_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700032089.rn.1 TC0700003968.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032083.rn.1 TC0700003968.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032082.rn.1 TC0700003968.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032088.rn.1 TC0700003968.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032085.rn.1 TC0700003968.rn.1 RTA TC
          Rat Expression Set 230 1376023_PM_at - -
          Rat Expression Set 230 1376023_at - -
          Rat Expression Set 230 1390771_at - -
          Rat Expression Set 230 1390771_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907574 - -
          Rat Genome U34 Set rc_AI010567_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22021792 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline