Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01638594_g1
          See other WNT4 GE Assays ›
          Gene Symbol:
          Wnt4
          Gene Name:
          wingless-type MMTV integration site family, member 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 155649238 - 155668065 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01638594_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wnt4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053402.2 NP_445854.1 4 - 5 594 62
          GenBank mRNA AF188608.1 - 4 - 5 595 62
          BC098752.1 - 4 - 5 615 7369699 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001721.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wnt4

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          branching involved in ureteric bud morphogenesis
          kidney development
          mesonephros development
          epithelial to mesenchymal transition
          embryonic epithelial tube formation
          liver development
          androgen biosynthetic process
          gamete generation
          sex differentiation
          male gonad development
          female gonad development
          cellular response to starvation
          negative regulation of gene expression
          negative regulation of steroid biosynthetic process
          Wnt signaling pathway
          protein palmitoylation
          regulation of cell-cell adhesion
          cell differentiation
          neuron differentiation
          female sex determination
          adrenal gland development
          negative regulation of cell migration
          positive regulation of bone mineralization
          positive regulation of aldosterone biosynthetic process
          response to estradiol
          positive regulation of collagen biosynthetic process
          T cell differentiation in thymus
          immature T cell proliferation in thymus
          tube morphogenesis
          non-canonical Wnt signaling pathway
          non-canonical Wnt signaling pathway via MAPK cascade
          negative regulation of fibroblast growth factor receptor signaling pathway
          hormone metabolic process
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of meiotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          oocyte development
          branching morphogenesis of an epithelial tube
          anatomical structure development
          smooth muscle cell differentiation
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          canonical Wnt signaling pathway
          somatotropin secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          mesenchymal to epithelial transition
          tertiary branching involved in mammary gland duct morphogenesis
          kidney morphogenesis
          negative regulation of wound healing
          mammary gland epithelium development
          positive regulation of dermatome development
          paramesonephric duct development
          negative regulation of testicular blood vessel morphogenesis
          cellular response to estradiol stimulus
          cellular response to hydrostatic pressure
          cellular response to transforming growth factor beta stimulus
          nephron development
          renal vesicle formation
          renal vesicle induction
          mesonephric tubule development
          metanephric tubule formation
          metanephric nephron development
          metanephric nephron morphogenesis
          establishment of protein localization to plasma membrane
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          negative regulation of male gonad development
          positive regulation of cortisol biosynthetic process
          negative regulation of testosterone biosynthetic process
          negative regulation of apoptotic signaling pathway
          molecular_function
          transcription corepressor activity
          frizzled binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P559414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41951 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P559414 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P559414 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P559414 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P559414 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28434 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10873023 - -
          Affymetrix Rat Gene 1.0 ST Array 10873025 - -
          Affymetrix Rat Gene 1.0 ST Array 10873022 - -
          Affymetrix Rat Gene 1.0 ST Array 10873024 - -
          Affymetrix Rat Gene 1.0 ST Array 10873026 - -
          Affymetrix Rat Gene 1.1 ST Array 10873021 - -
          Rat Genome 230 2.0 Array 1368641_PM_at - -
          Rat Genome 230 2.0 Array 1368641_at - -
          Affymetrix HT Rat Focus Array 1368641_at - -
          Affymetrix HT Rat Focus Array 1368641_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500012537.rn.1 TC0500001721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012536.rn.1 TC0500001721.rn.1 RTA TC
          Rat Expression Set 230 1368641_PM_at - -
          Rat Expression Set 230 1368641_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873021 - -
          Rat Genome U34 Set rc_AA900630_s_at - -
          Rat Genome U34 Set rc_AI172290_i_at - -
          Rat Genome U34 Set rc_AI103240_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21586566 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356435 - -
          RatRef-12 Expression BeadChip ILMN_64043 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline