Search
Search
| Gene Symbol: Gpr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080579.1 | NP_542146.1 | 1 - 2 | 128 | 84 | |
| XM_008763379.1 | XP_008761601.1 | 3 - 4 | 1044 | 84 | ||
| GenBank mRNA | BC089971.1 | - | 1 - 2 | 128 | 7315466 | 84 |
Set Membership: |
|