Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753606_g1
          See other SDCBP2 GE Assays ›
          Gene Symbol:
          Sdcbp2
          Gene Name:
          syndecan binding protein (syntenin) 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 147072133 - 147101917 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753606_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdcbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025692.1 NP_001020863.1 8 - 9 918 65
          XM_008762325.1 XP_008760547.1 6 - 7 2894 65
          XM_008762326.1 XP_008760548.1 9 - 10 1899 65
          GenBank mRNA BC099095.1 - 8 - 9 918 7459023 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002007.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdcbp2

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Process(es)

          protein C-terminus binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16766 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046552 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046552 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046552 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543769 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32500 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840759 - -
          Affymetrix Rat Gene 1.0 ST Array 10840751 - -
          Affymetrix Rat Gene 1.0 ST Array 10840752 - -
          Affymetrix Rat Gene 1.0 ST Array 10840754 - -
          Affymetrix Rat Gene 1.0 ST Array 10840753 - -
          Affymetrix Rat Gene 1.0 ST Array 10840755 - -
          Affymetrix Rat Gene 1.0 ST Array 10840757 - -
          Affymetrix Rat Gene 1.0 ST Array 10840756 - -
          Affymetrix Rat Gene 1.0 ST Array 10840758 - -
          Affymetrix Rat Gene 1.1 ST Array 10840750 - -
          Rat Genome 230 2.0 Array 1376973_PM_at - -
          Rat Genome 230 2.0 Array 1376973_at - -
          Affymetrix HT Rat Focus Array 1376973_PM_at - -
          Affymetrix HT Rat Focus Array 1376973_at - -
          Affymetrix Rat Clariom S Assay PSR0300014316.rn.1 TC0300002007.rn.1 RTA TC
          Rat Expression Set 230 1376973_at - -
          Rat Expression Set 230 1376973_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840750 - -
          Rat Genome U34 Set rc_AI044257_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21420770 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline