Search
Search
| Gene Symbol: Rnf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012195.1 | NP_001012195.1 | 3 - 4 | 426 | 61 | |
| XM_006240772.2 | XP_006240834.1 | 5 - 6 | 857 | 61 | ||
| XM_006240773.2 | XP_006240835.1 | 5 - 6 | 919 | 61 | ||
| XM_006240774.2 | XP_006240836.1 | 4 - 5 | 765 | 61 | ||
| XM_006240775.2 | XP_006240837.1 | 5 - 6 | 775 | 61 | ||
| XM_006240776.2 | XP_006240838.1 | 4 - 5 | 683 | 61 | ||
| XM_008765030.1 | XP_008763252.1 | 4 - 5 | 1776 | 61 | ||
| GenBank mRNA | BC083614.1 | - | 3 - 4 | 426 | 7133774 | 61 |
Set Membership: |
|