Search
Search
| Gene Symbol: Alg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108104.2 | NP_001101574.2 | 3 - 4 | 345 | 115 | |
| GenBank mRNA | BC161934.1 | - | 3 - 4 | 343 | 9022144 | 115 |
| FM050024.1 | - | 3 - 4 | 345 | 115 | ||
Set Membership: |
|