Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02346397_m1
          See other LDHD GE Assays ›
          Gene Symbol:
          Ldhd
          Gene Name:
          lactate dehydrogenase D
          Gene Aliases:
          Ac2-202
          Chromosome Location:
          Chr.19: 43826989 - 43848937 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02346397_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ldhd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008893.1 NP_001008893.1 10 - 11 1366 146
          GenBank mRNA AY321341.1 - 10 - 11 1366 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001475.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ldhd

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          UDP-N-acetylmuramate dehydrogenase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18171 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18171 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18171 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18171 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29069 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5759 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811171 - -
          Affymetrix Rat Gene 1.0 ST Array 10811174 - -
          Affymetrix Rat Gene 1.0 ST Array 10811173 - -
          Affymetrix Rat Gene 1.0 ST Array 10811172 - -
          Affymetrix Rat Gene 1.0 ST Array 10811164 - -
          Affymetrix Rat Gene 1.0 ST Array 10811167 - -
          Affymetrix Rat Gene 1.0 ST Array 10811176 - -
          Affymetrix Rat Gene 1.0 ST Array 10811161 - -
          Affymetrix Rat Gene 1.0 ST Array 10811163 - -
          Affymetrix Rat Gene 1.0 ST Array 10811170 - -
          Affymetrix Rat Gene 1.0 ST Array 10811166 - -
          Affymetrix Rat Gene 1.0 ST Array 10811168 - -
          Affymetrix Rat Gene 1.1 ST Array 10811160 - -
          Rat Genome 230 2.0 Array 1376746_at - -
          Rat Genome 230 2.0 Array 1382061_PM_at - -
          Rat Genome 230 2.0 Array 1376746_PM_at - -
          Rat Genome 230 2.0 Array 1382061_at - -
          Affymetrix HT Rat Focus Array 1376746_at - -
          Affymetrix HT Rat Focus Array 1382061_PM_at - -
          Affymetrix HT Rat Focus Array 1376746_PM_at - -
          Affymetrix HT Rat Focus Array 1382061_at - -
          Affymetrix Rat Clariom S Assay PSR1900012112.rn.1 TC1900001475.rn.1 RTA TC
          Rat Expression Set 230 1382061_at - -
          Rat Expression Set 230 1376746_PM_at - -
          Rat Expression Set 230 1382061_PM_at - -
          Rat Expression Set 230 1376746_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811160 - -
          Rat Genome U34 Set rc_AI639504_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358081 - -
          RatRef-12 Expression BeadChip ILMN_58624 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline