Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02348799_mH
          See other LY49S4 GE Assays ›
          Gene Symbol:
          Ly49s4
          Gene Name:
          Ly49 stimulatory receptor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 164470396 - 164493252 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02348799_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly49s4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009487.1 NP_001009487.1 325 140
          GenBank mRNA AY649831.1 - 325 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003883.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly49s4

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142727 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75699 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90672 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866171 - -
          Affymetrix Rat Gene 1.0 ST Array 10866177 - -
          Affymetrix Rat Gene 1.0 ST Array 10866170 - -
          Affymetrix Rat Gene 1.0 ST Array 10866168 - -
          Affymetrix Rat Gene 1.0 ST Array 10866169 - -
          Affymetrix Rat Gene 1.0 ST Array 10866165 - -
          Affymetrix Rat Gene 1.0 ST Array 10866164 - -
          Affymetrix Rat Gene 1.0 ST Array 10866175 - -
          Affymetrix Rat Gene 1.1 ST Array 10866163 - -
          Affymetrix Rat Gene 1.1 ST Array 10866167 - -
          Affymetrix Rat Clariom S Assay PSR0400027722.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027723.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027726.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027724.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027725.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027720.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027727.rn.1 TC0400003883.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866167 - -
          Affymetrix Rat Gene 1.0 ST Array 10866163 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650238 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline