Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02396766_m1
          See other CPLX1 GE Assays ›
          Gene Symbol:
          Cplx1
          Gene Name:
          complexin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 2194891 - 2226610 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02396766_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cplx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022864.3 NP_074055.1 2 - 3 280 73
          XM_006250571.2 XP_006250633.1 3 - 4 406 73
          GenBank mRNA BC093605.1 - 2 - 3 283 7317504 73
          D70817.1 - 3 - 4 315 73
          U35098.1 - 3 - 4 313 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000035.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cplx1

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptic vesicle exocytosis
          insulin secretion
          SNARE binding
          neurotransmitter transporter activity
          syntaxin-1 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072878 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86484 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57264 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072873 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072873 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072873 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072878 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P413294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90291 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9334 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770982 - -
          Affymetrix Rat Gene 1.0 ST Array 10770983 - -
          Affymetrix Rat Gene 1.0 ST Array 10770978 - -
          Affymetrix Rat Gene 1.0 ST Array 10770981 - -
          Affymetrix Rat Gene 1.0 ST Array 10770980 - -
          Affymetrix Rat Gene 1.0 ST Array 10770979 - -
          Affymetrix Rat Gene 1.1 ST Array 10770976 - -
          Rat Genome 230 2.0 Array 1369811_PM_at - -
          Rat Genome 230 2.0 Array 1369811_at - -
          Rat Genome 230 2.0 Array 1398357_PM_at - -
          Rat Genome 230 2.0 Array 1398357_at - -
          Affymetrix HT Rat Focus Array 1398357_PM_at - -
          Affymetrix HT Rat Focus Array 1369811_at - -
          Affymetrix HT Rat Focus Array 1398357_at - -
          Affymetrix HT Rat Focus Array 1369811_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400000292.rn.1 TC1400000035.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000291.rn.1 TC1400000035.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400000290.rn.1 TC1400000035.rn.1 RTA TC
          Rat Expression Set 230 1398357_PM_at - -
          Rat Expression Set 230 1369811_PM_at - -
          Rat Expression Set 230 1398357_at - -
          Rat Expression Set 230 1369811_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770976 - -
          Rat Genome U34 Set D70817_at - -
          Rat Genome U34 Set rc_AA943896_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20905752 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039998 - -
          RatRef-12 Expression BeadChip ILMN_68168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline