Search
Search
| Gene Symbol: Ptbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271057.1 | NP_001257986.1 | 4 - 5 | 495 | 88 | |
| NM_022516.5 | NP_071961.2 | 4 - 5 | 495 | 88 | ||
| GenBank mRNA | BC061858.1 | - | 4 - 5 | 490 | 5598495 | 88 |
| CK365398.1 | - | 1 - 2 | 81 | 88 | ||
| FN803753.1 | - | 4 - 5 | 465 | 88 | ||
| X60789.1 | - | 4 - 5 | 458 | 88 | ||
| X74565.1 | - | 4 - 5 | 476 | 88 | ||
Set Membership: |
|