Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn99999185_m1
          See other IL18 GE Assays ›
          Gene Symbol:
          Il18
          Gene Name:
          interleukin 18
          Gene Aliases:
          IL-18
          Chromosome Location:
          Chr.8: 55009666 - 55016286 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn99999185_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019165.1 NP_062038.1 3 - 4 225 107
          GenBank mRNA AJ222813.1 - 3 - 4 225 107
          AY077842.1 - 453 107
          AY258448.1 - 3 - 4 225 107
          U77776.1 - 3 - 4 225 107
          U77777.1 - 3 - 4 225 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003653.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il18

          Panther Classification:

          Molecular Function -

          cytokine interleukin superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          osteoblast differentiation
          response to hypoxia
          inflammatory response
          learning or memory
          response to radiation
          response to cold
          positive regulation of gene expression
          positive regulation of smooth muscle cell migration
          natural killer cell activation
          regulation of cell adhesion
          lung development
          sleep
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-17 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of natural killer cell proliferation
          positive regulation of superoxide anion generation
          positive regulation of collagen biosynthetic process
          negative regulation of neutrophil apoptotic process
          T-helper 1 type immune response
          interferon-gamma biosynthetic process
          positive regulation of activated T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          positive regulation of vascular permeability
          response to peptide hormone
          positive regulation of neuron apoptotic process
          regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of myoblast differentiation
          digestive tract morphogenesis
          positive regulation of smooth muscle cell proliferation
          negative regulation of peptidyl-tyrosine phosphorylation
          detection of mechanical stimulus involved in sensory perception of pain
          positive regulation of NK T cell proliferation
          response to hyperoxia
          cellular response to hydrogen peroxide
          cellular response to lipopolysaccharide
          cellular response to mechanical stimulus
          cellular response to cAMP
          cellular response to interferon-gamma
          cellular response to tumor necrosis factor
          cellular response to parathyroid hormone stimulus
          cellular response to peptide hormone stimulus
          cellular response to organic cyclic compound
          positive regulation of male germ cell proliferation
          positive regulation of blood vessel remodeling
          cytokine activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline