Search
Search

ctdsp2| Gene Symbol: ctdsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006792.1 | NP_001006793.1 | 3 - 4 | 637 | 87 | |
| XM_012956751.2 | XP_012812205.1 | 4 - 5 | 1099 | 87 | ||
| GenBank mRNA | BC076658.1 | - | 3 - 4 | 637 | 87 | |
Set Membership: |
|