Search
Search

ocln| Gene Symbol: ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015948.1 | NP_001015948.1 | 5 - 6 | 1180 | 76 | |
| GenBank mRNA | BC123964.1 | - | 5 - 6 | 1173 | 76 | |
| CR761445.2 | - | 5 - 6 | 1180 | 76 | ||
Set Membership: |
|